Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ASB18
Entrez Gene ID
101103518
Gene Name
ankyrin repeat and SOCS box containing 18
Gene Aliases
-
Chromosome Location
Chr.1: 4874137 - 4951760 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004001771.4 XP_004001820.2 5 - 6 1257 77

View Full Product Details