Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FADS1
Entrez Gene ID
101109049
Gene Name
fatty acid desaturase 1
Gene Aliases
-
Chromosome Location
Chr.21: 42128559 - 42142186 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019593.4 XP_004019642.1 8 - 9 1093 71
XM_012102002.3 XP_011957392.1 8 - 9 899 71
XM_027959823.1 XP_027815624.1 8 - 9 937 71

View Full Product Details