Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC10A4
Entrez Gene ID
101113071
Gene Name
solute carrier family 10 member 4
Gene Aliases
-
Chromosome Location
Chr.6: 73757274 - 73763233 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010062.4 XP_004010111.3 1 - 2 703 63

View Full Product Details