Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INCA1
Entrez Gene ID
101107802
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 36948348 - 36956489 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012597.4 XP_004012646.1 4 - 5 386 60
XM_012185566.3 XP_012040956.1 4 - 5 1049 60
XM_012185577.2 XP_012040967.1 4 - 5 1691 60
XM_015098601.2 XP_014954087.1 3 - 4 1595 60
XM_015098603.2 XP_014954089.1 4 - 5 1996 60
XM_027975075.1 XP_027830876.1 4 - 5 734 60
XM_027975076.1 XP_027830877.1 3 - 4 2080 60
XM_027975077.1 XP_027830878.1 4 - 5 1990 60
XM_027975078.1 XP_027830879.1 3 - 4 1542 60

View Full Product Details