Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMB
Entrez Gene ID
101108605
Gene Name
neuromedin B
Gene Aliases
-
Chromosome Location
Chr.18: 19730711 - 19733994 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017766.4 XP_004017815.1 2 - 3 376 74

View Full Product Details