Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC8A2
Entrez Gene ID
101102171
Gene Name
solute carrier family 8 member A2
Gene Aliases
-
Chromosome Location
Chr.14: 57875881 - 57909821 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027978539.1 XP_027834340.1 8 - 9 2263 73

View Full Product Details