Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL1F10
Entrez Gene ID
101111084
Gene Name
interleukin 1 family member 10
Gene Aliases
-
Chromosome Location
Chr.3: 63928033 - 63931931 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027965786.1 XP_027821587.1 3 - 4 274 122

View Full Product Details