Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS18
Entrez Gene ID
101111552
Gene Name
VPS18 core subunit of CORVET and HOPS complexes
Gene Aliases
-
Chromosome Location
Chr.7: 36422359 - 36431144 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004010456.4 XP_004010505.3 1 - 2 449 128

View Full Product Details