Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDHAF2
Entrez Gene ID
101107829
Gene Name
succinate dehydrogenase complex assembly factor 2
Gene Aliases
-
Chromosome Location
Chr.21: 41804169 - 41813708 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019588.3 XP_004019637.1 3 - 4 479 107

View Full Product Details