Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB32
Entrez Gene ID
101106354
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
-
Chromosome Location
Chr.14: 49154982 - 49161269 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027977451.1 XP_027833252.1 2 - 3 4397 109

View Full Product Details