Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK2A
Entrez Gene ID
101105549
Gene Name
divergent protein kinase domain 2A
Gene Aliases
C1H3orf58
Chromosome Location
Chr.1: 267541514 - 267563249 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959602.1 XP_027815403.1 2 - 3 1019 96

View Full Product Details