Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD5
Entrez Gene ID
101113440
Gene Name
sushi domain containing 5
Gene Aliases
-
Chromosome Location
Chr.19: 8098226 - 8144600 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018573.4 XP_004018622.2 4 - 5 897 93

View Full Product Details