Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF3C4
Entrez Gene ID
101119386
Gene Name
general transcription factor IIIC subunit 4
Gene Aliases
-
Chromosome Location
Chr.3: 4348774 - 4364896 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015094120.2 XP_014949606.2 3 - 4 2313 106

View Full Product Details