Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SYCN
Entrez Gene ID
101115647
Gene Name
syncollin
Gene Aliases
-
Chromosome Location
Chr.14: 51824101 - 51824559 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100209.2 XP_014955695.2 1 - 1 216 114

View Full Product Details