Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STK17A
Entrez Gene ID
101119390
Gene Name
serine/threonine kinase 17a
Gene Aliases
-
Chromosome Location
Chr.4: 85042338 - 85080283 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968745.1 XP_027824546.1 2 - 3 539 107

View Full Product Details