Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LIX1L
Entrez Gene ID
101118146
Gene Name
limb and CNS expressed 1 like
Gene Aliases
-
Chromosome Location
Chr.1: 104968390 - 104991682 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976361.1 XP_027832162.1 2 - 3 448 93

View Full Product Details