Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM241A
Entrez Gene ID
114115388
Gene Name
family with sequence similarity 241 member A
Gene Aliases
C6H4orf32
Chromosome Location
Chr.6: 16487446 - 16503179 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027971375.1 XP_027827176.1 3 - 3 376 89

View Full Product Details