Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP4
Entrez Gene ID
100008691
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
TIMP-4
Chromosome Location
Chr.9: 11767817 - 11773763 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195690.1 NP_001182619.1 2 - 3 241 112
GenBank mRNA AF069715.1 - 1 - 2 89 112

View Full Product Details