Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH11
Entrez Gene ID
100008870
Gene Name
dynein axonemal heavy chain 11
Gene Aliases
DNAHC11
Chromosome Location
Chr.10: 6670834 - 7032479 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008261514.2 XP_008259736.1 61 - 62 10191 86
GenBank mRNA AJ920326.1 - 4 - 5 500 86

View Full Product Details