Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INSR
Entrez Gene ID
100008911
Gene Name
insulin receptor
Gene Aliases
DAF-2
Chromosome Location
Chr.Unknown: 1077774 - 1214036 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008249177.2 XP_008247399.1 - 1675 76
XM_017337973.1 XP_017193462.1 - 1675 76
XM_017337974.1 XP_017193463.1 - 1675 76
XM_017337975.1 XP_017193464.1 - 1675 76
GenBank mRNA AY339877.1 - - 1531 76

View Full Product Details