Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDGFD
Entrez Gene ID
100008912
Gene Name
platelet derived growth factor D
Gene Aliases
PDGF-D
Chromosome Location
Chr.1: 112364811 - 112640557 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195762.1 NP_001182691.1 6 - 7 987 84
XM_002708534.3 XP_002708580.1 6 - 7 1397 84
GenBank mRNA AY347260.1 - 5 - 6 819 84

View Full Product Details