Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC4A9
Entrez Gene ID
100008621
Gene Name
solute carrier family 4 member 9
Gene Aliases
AE4A
Chromosome Location
Chr.3: 22495679 - 22534757 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082005.1 NP_001075474.1 20 - 21 2836 110
XM_008254828.2 XP_008253050.1 20 - 21 3386 110
XM_008254829.2 XP_008253051.1 20 - 21 3333 110
XM_017340967.1 XP_017196456.1 20 - 21 3543 110
XM_017340968.1 XP_017196457.1 20 - 21 3544 110
XM_017340969.1 XP_017196458.1 20 - 21 3495 110
XM_017340970.1 XP_017196459.1 20 - 21 3475 110
XM_017340971.1 XP_017196460.1 19 - 20 3429 110
XM_017340972.1 XP_017196461.1 20 - 21 3427 110
XM_017340973.1 XP_017196462.1 20 - 21 3390 110
XM_017340974.1 XP_017196463.1 19 - 20 2699 110
GenBank mRNA AB038263.1 - 20 - 21 2836 110
AB038264.1 - 20 - 21 2788 110

View Full Product Details