Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NDEL1
Entrez Gene ID
100008657
Gene Name
nudE neurodevelopment protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.19: 10830833 - 10863389 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082020.1 NP_001075489.1 5 - 6 872 75
GenBank mRNA AF015037.3 - 5 - 6 872 75

View Full Product Details