Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A1
Entrez Gene ID
100008659
Gene Name
solute carrier family 22 member 1
Gene Aliases
OCT1, RBOCT1
Chromosome Location
Chr.12: 149873910 - 149896692 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082022.1 NP_001075491.1 7 - 8 1330 105
GenBank mRNA AF015958.1 - 7 - 8 1330 105

View Full Product Details