Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GYG1
Entrez Gene ID
100009058
Gene Name
glycogenin 1
Gene Aliases
GYG
Chromosome Location
Chr.14: 42793516 - 42828602 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082241.1 NP_001075710.1 6 - 7 685 127
GenBank mRNA L01791.1 - 6 - 7 685 127

View Full Product Details