Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSE
Entrez Gene ID
100009063
Gene Name
cathepsin E
Gene Aliases
-
Chromosome Location
Chr.16: 66242101 - 66257568 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082244.1 NP_001075713.1 8 - 9 1072 55
XM_017347578.1 XP_017203067.1 9 - 10 1177 55
GenBank mRNA L08418.1 - 8 - 9 1072 55

View Full Product Details