Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHBG
Entrez Gene ID
100009224
Gene Name
sex hormone binding globulin
Gene Aliases
-
Chromosome Location
Chr.19: 11560507 - 11563050 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082370.1 NP_001075839.1 5 - 6 758 62
GenBank mRNA U57553.1 - 5 - 6 724 62
X12661.1 - - 400 62

View Full Product Details