Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC15A2
Entrez Gene ID
100009433
Gene Name
solute carrier family 15 member 2
Gene Aliases
OPT-3
Chromosome Location
Chr.14: 97075969 - 97116468 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082700.1 NP_001076169.1 9 - 10 880 75
XM_017346648.1 XP_017202137.1 8 - 9 847 75
GenBank mRNA U32507.1 - 9 - 10 880 75

View Full Product Details