Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD1C
Entrez Gene ID
100009419
Gene Name
CD1c molecule
Gene Aliases
-
Chromosome Location
Chr.13: 34954465 - 34958093 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082689.1 NP_001076158.1 2 - 3 324 156
GenBank mRNA AF276978.1 - 2 - 3 324 156

View Full Product Details