Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRO
Entrez Gene ID
100009529
Gene Name
protein tyrosine phosphatase receptor type O
Gene Aliases
GLEPP1, PTP
Chromosome Location
Chr.8: 23969041 - 24227954 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082750.1 NP_001076219.1 13 - 14 2483 111
NM_001170980.1 NP_001164451.1 1 - 2 265 111
XM_008259342.2 XP_008257564.1 13 - 14 2522 111
XM_017343246.1 XP_017198735.1 13 - 14 2435 111
GenBank mRNA U09490.1 - 13 - 14 2483 111
U32587.1 - 1 - 2 265 111

View Full Product Details