Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2CB
Entrez Gene ID
100009300
Gene Name
protein phosphatase 2 catalytic subunit beta
Gene Aliases
-
Chromosome Location
Chr.2: 76926974 - 76957892 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101707.1 NP_001095177.1 2 - 3 311 107
GenBank mRNA Y00763.1 - - 311 107

View Full Product Details