Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RS1
Entrez Gene ID
100126075
Gene Name
retinoschisin 1
Gene Aliases
-
Chromosome Location
Chr.X: 4481879 - 4501540 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109823.1 NP_001103293.1 4 - 5 331 60
GenBank mRNA EU074985.1 - 4 - 5 331 60

View Full Product Details