Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FTL
Entrez Gene ID
100009254
Gene Name
ferritin light chain
Gene Aliases
FTL2
Chromosome Location
Chr.Unknown: 86633 - 87941 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101688.1 NP_001095158.1 - 248 67
GenBank mRNA X07830.1 - - 248 67

View Full Product Details