Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT2A1
Entrez Gene ID
100008598
Gene Name
sulfotransferase family 2A member 1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 124783 - 143363 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081987.1 NP_001075456.1 - 753 74
GenBank mRNA AB006053.1 - - 753 74

View Full Product Details