Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100328613
Entrez Gene ID
100328613
Gene Name
polycyclic hydrocarbon-inducible cytochrome P450c
Gene Aliases
CYP1A1, CYPIA1
Chromosome Location
Chr.Unknown: 237273 - 242649 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171072.1 NP_001164543.1 - 1058 76
XM_008252400.2 XP_008250622.1 - 1071 76
GenBank mRNA M11727.1 - - 747 76
M19917.1 - - 798 76
X05685.1 - - 1058 76

View Full Product Details