Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OSTM1
Entrez Gene ID
100328697
Gene Name
osteoclastogenesis associated transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.12: 96059590 - 96090626 on Build OryCun2.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171322.1 NP_001164793.1 4 - 5 771 70

View Full Product Details