Gene Transcripts

Gene Symbol
Clmp
Entrez Gene ID
286939
Gene Name
CXADR-like membrane protein
Gene Aliases
ACAM, Asam, Ol16
Chromosome Location
Chr. 8: 44847157 - 44975460 on Build Rnor_6.0
UniGene ID
Rn.163013
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173154.1 NP_775177.1 7 1095
GenBank mRNA AF302047.1 - 7 1095

View Full Product Details