Gene Transcripts

Gene Symbol
Appl1
Entrez Gene ID
290537
Gene Name
adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
Gene Aliases
RGD1309388
Chromosome Location
Chr. 16: 2560103 - 2602035 on Build Rnor_6.0
UniGene ID
Rn.224576
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771023.1 XP_008769245.1 9 916
GenBank mRNA FQ226179.1 - 9 916

View Full Product Details