Gene Transcripts

Gene Symbol
Urb2
Entrez Gene ID
292087
Gene Name
URB2 ribosome biogenesis 2 homolog (S. cerevisiae)
Gene Aliases
RGD1307980
Chromosome Location
Chr. 19: 56803708 - 56827017 on Build Rnor_6.0
UniGene ID
Rn.6429
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135708.1 NP_001129180.1 3 995

View Full Product Details