Gene Transcripts

Gene Symbol
Ltbp4
Entrez Gene ID
292734
Gene Name
latent transforming growth factor beta binding protein 4
Gene Aliases
Chromosome Location
Chr. 1: 84118046 - 84150084 on Build Rnor_6.0
UniGene ID
Rn.7961
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170336.1 NP_001163807.1 10 1263
XM_006228570.2 XP_006228632.1 7 1105
XM_008759116.1 XP_008757338.1 10 1170

View Full Product Details