Gene Transcripts

Gene Symbol
Depdc1
Entrez Gene ID
295538
Gene Name
DEP domain containing 1
Gene Aliases
Depdc1a
Chromosome Location
Chr. 2: 264704734 - 264738265 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233537.2 XP_006233599.1 7 982
XM_227798.8 XP_227798.5 7 984

View Full Product Details