Gene Transcripts

Gene Symbol
Sh2d7
Entrez Gene ID
300718
Gene Name
SH2 domain containing 7
Gene Aliases
RGD1309791
Chromosome Location
Chr. 8: 59110859 - 59122584 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008766361.1 XP_008764583.1 5 1604
XM_008766362.1 XP_008764584.1 5 1604

View Full Product Details