Gene Transcripts

Gene Symbol
Supt6h
Entrez Gene ID
303281
Gene Name
suppressor of Ty 6 homolog (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 65470470 - 65507581 on Build Rnor_6.0
UniGene ID
Rn.7144
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191820.1 NP_001178749.1 26 3808
XM_008768006.1 XP_008766228.1 26 3469
XM_008768007.1 XP_008766229.1 27 3660
XM_008768008.1 XP_008766230.1 26 3660

View Full Product Details