Gene Transcripts

Gene Symbol
Adcy1
Entrez Gene ID
305509
Gene Name
adenylate cyclase 1 (brain)
Gene Aliases
Ac1
Chromosome Location
Chr. 14: 87311912 - 87424511 on Build Rnor_6.0
UniGene ID
Rn.214145
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107239.1 NP_001100709.1 4 786
XM_008770320.1 XP_008768542.1 6 1016
XM_008770321.1 XP_008768543.1 5 944
XM_008770322.1 XP_008768544.1 4 365

View Full Product Details