Gene Transcripts

Gene Symbol
Samd4a
Entrez Gene ID
305826
Gene Name
sterile alpha motif domain containing 4A
Gene Aliases
Samd4
Chromosome Location
Chr. 15: 23777000 - 23879067 on Build Rnor_6.0
UniGene ID
Rn.224832
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107254.1 NP_001100724.1 3 1071
XM_008770515.1 XP_008768737.1 3 1909
GenBank mRNA BC168894.1 - 3 1053

View Full Product Details