Gene Transcripts

Gene Symbol
Ano5
Entrez Gene ID
308637
Gene Name
anoctamin 5
Gene Aliases
RGD1564265
Chromosome Location
Chr. 1: 106873578 - 106971769 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229266.2 XP_006229328.1 7 1754
XM_006229268.2 XP_006229330.1 7 1753

View Full Product Details