Gene Transcripts

Gene Symbol
Ddb2
Entrez Gene ID
100362121
Gene Name
damage specific DNA binding protein 2
Gene Aliases
Chromosome Location
Chr. 3: 80030437 - 80052986 on Build Rnor_6.0
UniGene ID
Rn.154614
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271346.1 NP_001258275.1 4 713
XM_006234498.2 XP_006234560.1 4 706
XM_006234499.2 XP_006234561.1 4 746

View Full Product Details