Gene Transcripts

Gene Symbol
Clec2h
Entrez Gene ID
312745
Gene Name
C-type lectin domain family 2, member H
Gene Aliases
Clecsf2
Chromosome Location
Chr. 4: 162810384 - 162844603 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008763345.1 XP_008761567.1 2 1480

View Full Product Details