Gene Transcripts

Gene Symbol
Dos
Entrez Gene ID
314622
Gene Name
downstream of Stk11
Gene Aliases
Chromosome Location
Chr. 7: 12433422 - 12441048 on Build Rnor_6.0
UniGene ID
Rn.91597
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108070.1 NP_001101540.1 10 1541
GenBank mRNA BC169056.1 - 1 153

View Full Product Details