Gene Transcripts

Gene Symbol
Skor1
Entrez Gene ID
315748
Gene Name
SKI family transcriptional corepressor 1
Gene Aliases
Lbxcor1, RGD1307687
Chromosome Location
Chr. 8: 68029930 - 68040504 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006243246.2 XP_006243308.2 7, 8 2767
XM_008766381.1 XP_008764603.1 7, 8 2646

View Full Product Details